missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRAME Antibody (CL5146), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61148
This item is not returnable.
View return policy
Description
PRAME Monoclonal specifically detects PRAME in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| PRAME | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| PRAME | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:LLLSHIHASSYISPEKEEQYIAQFTSQFLSLQCLQALYVDSLFFLRGRLDQLLRHVMNPLETLSITNCRLSEGDVMHLSQSPSVSQLSVLSLSGVMLTDVSPEPLQALLERASATLQDLVFDECGI | |
| 100 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| CL5146 | |
| Western Blot 1 μg/mL, Immunocytochemistry/Immunofluorescence 2-10 μg/mL | |
| cancer/testis antigen 130, MAPEOIP4CT130, melanoma antigen preferentially expressed in tumors, OIP-4, Opa-interacting protein 4, Opa-interacting protein OIP4, preferentially expressed antigen in melanoma, Preferentially expressed antigen of melanoma | |
| Mouse | |
| Protein A purified | |
| Cancer | |
| 23532 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction