missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRAME Antibody (CL5146), Novus Biologicals™
Mouse Monoclonal Antibody
€ 369.00 - € 564.00
Specifications
| Antigen | PRAME |
|---|---|
| Clone | CL5146 |
| Dilution | Western Blot 1 μg/mL, Immunocytochemistry/Immunofluorescence 2-10 μg/mL |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18037323
|
Novus Biologicals
NBP2-61148 |
100 μL |
€ 564.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18613358
|
Novus Biologicals
NBP2-61148-25ul |
25 μL |
€ 369.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PRAME Monoclonal specifically detects PRAME in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| PRAME | |
| Western Blot 1 μg/mL, Immunocytochemistry/Immunofluorescence 2-10 μg/mL | |
| Monoclonal | |
| Purified | |
| Cancer | |
| 23532 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:LLLSHIHASSYISPEKEEQYIAQFTSQFLSLQCLQALYVDSLFFLRGRLDQLLRHVMNPLETLSITNCRLSEGDVMHLSQSPSVSQLSVLSLSGVMLTDVSPEPLQALLERASATLQDLVFDECGI | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| CL5146 | |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Human | |
| PRAME | |
| IgG2a | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title