Learn More
VELP Scientifica™ Preventative Maintenance Full Kit for CSF6 Filtration Unit
Shop All Velp Products
Click to view available options
For Use With (Equipment):
CSF6 Filtration Unit
Unit Size:
Each
Description
The PM full Kit is designed to keep you running with high performance while ensuring long life and reliability, even under high workloads.
- Reduces unplanned downtime with consequent improvement of profit and production performance
- Lowers costs for the purchase of necessary spare parts and eventual extraordinary maintenance
- Reduces lost time associated with unplanned service intervals and instrument downtime
- Includes all necessary spare parts from the manufacturer
- Preventative maintenance needs to be carried out by a VELP™ authorized technician to guarantee durability, and performance per VELP™ quality standards
Specifications
Specifications
| For Use With (Equipment) | CSF6 Filtration Unit |
| Product Type | Maintenance Kit |
| Includes | 18 x EPDM CSF6 COEX Gaskets, 12 x OR 4131 FKM, 10 x T-Piece Union (6 mm Dia.), 5 x L-Piece Union (6 mm Dia.), 6 x Disk Wire Mesh, Norprene Tube (7.9 x 11.1 mm Dia.), Set of CSF6 Tubes |