missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRL-3/PTP4A3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93734-0.1ml
This item is not returnable.
View return policy
Description
PRL-3/PTP4A3 Polyclonal antibody specifically detects PRL-3/PTP4A3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| PRL-3/PTP4A3 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin | |
| PRL3EC 3.1.3.48, PRL-3potentially prenylated protein tyrosine phosphatase, PRL-Rprotein tyrosine phosphatase type IVA 3, protein tyrosine phosphatase type IVA, member 3, Protein-tyrosine phosphatase 4a3, Protein-tyrosine phosphatase of regenerating liver 3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-148 of human PTP4A3 (NP_009010.2). MARMNRPAPVEVSYKHMRFLITHNPTNATLSTFIEDLKKYGATTVVRVCEVTYDKTPLEKDGITVVDWPFDDGAPPPGKVVEDWLSLVKAKFCEAPGSCVAVHCVAGLGRKRRGAINSKQLTYLEKYRPKQRLRFKDPHTHKTRCCVM | |
| 0.1 mL | |
| Protein Phosphatase | |
| 11156 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction