missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRL-3/PTP4A3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 201.00 - € 470.00
Specifications
| Antigen | PRL-3/PTP4A3 |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18613111
|
Novus Biologicals
NBP2-93734-0.02ml |
0.02 mL |
€ 201.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18613431
|
Novus Biologicals
NBP2-93734-0.1ml |
0.1 mL |
€ 470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PRL-3/PTP4A3 Polyclonal antibody specifically detects PRL-3/PTP4A3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| PRL-3/PTP4A3 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Protein Phosphatase | |
| PBS (pH 7.3), 50% glycerol | |
| 11156 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:1000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| PRL3EC 3.1.3.48, PRL-3potentially prenylated protein tyrosine phosphatase, PRL-Rprotein tyrosine phosphatase type IVA 3, protein tyrosine phosphatase type IVA, member 3, Protein-tyrosine phosphatase 4a3, Protein-tyrosine phosphatase of regenerating liver 3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-148 of human PTP4A3 (NP_009010.2). MARMNRPAPVEVSYKHMRFLITHNPTNATLSTFIEDLKKYGATTVVRVCEVTYDKTPLEKDGITVVDWPFDDGAPPPGKVVEDWLSLVKAKFCEAPGSCVAVHCVAGLGRKRRGAINSKQLTYLEKYRPKQRLRFKDPHTHKTRCCVM | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title