missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRMT7 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33258-20ul
This item is not returnable.
View return policy
Description
PRMT7 Monoclonal antibody specifically detects PRMT7 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| PRMT7 | |
| Monoclonal | |
| Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL | |
| [Myelin basic protein]-arginine N-methyltransferase PRMT7, EC 2.1.1, EC 2.1.1.125, EC 2.1.1.126, FLJ10640, Histone-arginine N-methyltransferase PRMT7, KIAA1933EC 2.1.1.-, myelin basic protein-arginine N-methyltransferase, protein arginine methyltransferase 7, protein arginine N-methyltransferase 7 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human PRMT7 (Q9NVM4).,, Sequence:, MKIFCSRANPTTGSVEWLEEDEHYDYHQEIARSSYADMLHDKDRNVKYYQGIRAAVSRVKDRGQKALVLDIGTGTGLLSMMAVTAGADFCYAIEVFKPMA | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 54496 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction