Learn More
Velp Scientifica™ Product Manual
Product Code. 11765654 Shop All Velp Products
Change view
Click to view available options
For Use With (Equipment):
COEX
CSF6
DKL Series
FIWE Series Raw Fiber Extractors
GDE
HU6
JP
NDA 701 and 702
OXITEST
SER 148 Series Solvent Extractors
SMS
Unit Size:
Each
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
11765654 DKL Series Each
15309584 NDA 701 and 702 Each
15309624 COEX Each
15389614 CSF6 Each
15399614 GDE Each
15319624 HU6 Each
15359554 SER 148 Series Solvent Extractors Each
15369554 FIWE Series Raw Fiber Extractors Each
15339624 JP Each
15329624 SMS Each
15349614 OXITEST Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Product Code. 11765654 Supplier Velp Scientifica™ Supplier No. A00000186
Only null left
Add to basket
Add to basket
Velp Scientifica™ Product Manual are for use with a wide range of products.
  • Inlcudes one manual for the desiered product
  • For use with a wide range of Velp Scientifica™ products
Specifications
Format Manual
For Use With (Equipment) DKL Series
Product Type Operating Instructions
Quantity 1/Ea.