missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Profilin 1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11680-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a ubiquitous actin monomer-binding protein belonging to the profilin family. It is thought to regulate actin polymerization in response to extracellular signals. Deletion of this gene is associated with Miller-Dieker syndrome.Specifications
| Profilin 1 | |
| Polyclonal | |
| Unconjugated | |
| PFN1 | |
| Epididymis tissue protein Li 184a, PFN1, Profilin 1, Profilin I | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 18643, 5216, 64303 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P07737, P62962, P62963 | |
| PFN1 | |
| Profilin 1 Fusion Protein Ag2286 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction