missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Progesterone R/NR3C3 Antibody (PR501), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-33368-20ug
This item is not returnable.
View return policy
Description
Progesterone R/NR3C3 Monoclonal specifically detects Progesterone R/NR3C3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Progesterone R/NR3C3 | |
| Monoclonal | |
| 0.2mg/mL | |
| Western Blot 0.5-1ug/ml, Immunohistochemistry-Paraffin 0.5-1ug/ml, SDS-Page | |
| P06401 | |
| PGR | |
| A progesterone receptor recombinant protein was used as the immunogen for the antibody. The specific epitope has not been mapped. | |
| 20 μg | |
| Breast Cancer, Cancer | |
| 5241 | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry (Paraffin), SDS-Page | |
| PR501 | |
| Unconjugated | |
| PBS with 0.05% BSA. with 0.05% Sodium Azide | |
| NR3C3PRNuclear receptor subfamily 3 group C member 3, progesterone receptor | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| This MAb is specific to progesterone receptor and shows minimal cross-reaction with other members of the family. Progesterone receptor is expressed as two major isoforms, PR-A (81kDa) and PR-B (116kDa). Expression of PgR has been suggested to reflect a intact estrogen regulatory machinery and therefore, predict better clinical response to endocrine therapy than ER alone. | |
| Store at 4C. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction