missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Progesterone R/NR3C3 Antibody (PR501), Novus Biologicals™
Mouse Monoclonal Antibody
€ 264.00 - € 474.00
Specifications
| Antigen | Progesterone R/NR3C3 |
|---|---|
| Clone | PR501 |
| Concentration | 0.2mg/mL |
| Dilution | Western Blot 0.5-1ug/ml, Immunohistochemistry-Paraffin 0.5-1ug/ml, SDS-Page |
| Applications | Western Blot, Immunohistochemistry (Paraffin), SDS-Page |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18162373
|
Novus Biologicals
NBP2-33368-0.1MG |
0.1 mg |
€ 474.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18463022
|
Novus Biologicals
NBP2-33368-20ug |
20 μg |
€ 264.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Progesterone R/NR3C3 Monoclonal specifically detects Progesterone R/NR3C3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Progesterone R/NR3C3 | |
| 0.2mg/mL | |
| Western Blot, Immunohistochemistry (Paraffin), SDS-Page | |
| Unconjugated | |
| Mouse | |
| Breast Cancer, Cancer | |
| PBS with 0.05% BSA. with 0.05% Sodium Azide | |
| NR3C3PRNuclear receptor subfamily 3 group C member 3, progesterone receptor | |
| PGR | |
| IgG1 κ | |
| Protein G purified | |
| This MAb is specific to progesterone receptor and shows minimal cross-reaction with other members of the family. Progesterone receptor is expressed as two major isoforms, PR-A (81kDa) and PR-B (116kDa). Expression of PgR has been suggested to reflect a intact estrogen regulatory machinery and therefore, predict better clinical response to endocrine therapy than ER alone. |
| PR501 | |
| Western Blot 0.5-1ug/ml, Immunohistochemistry-Paraffin 0.5-1ug/ml, SDS-Page | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P06401 | |
| 5241 | |
| A progesterone receptor recombinant protein was used as the immunogen for the antibody. The specific epitope has not been mapped. | |
| Primary | |
| Store at 4C. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title