missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteinase 3/Myeloblastin/PRTN3 Antibody (3B4), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-51573-0.025ml
This item is not returnable.
View return policy
Description
Proteinase 3/Myeloblastin/PRTN3 Monoclonal specifically detects Proteinase 3/Myeloblastin/PRTN3 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| Proteinase 3/Myeloblastin/PRTN3 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, ELISA 1:10000 | |
| PRTN3 | |
| Purified recombinant fragment of human Proteinase 3 expressed in E. Coli. | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| 3B4 | |
| Unconjugated | |
| ACPA, AGP7PR3, azurophil granule protein 7, C-ANCA, EC 3.4.21, EC 3.4.21.76, Leukocyte proteinase 3, MBN, MBT, myeloblastin, Neutrophil proteinase 4, NP4, NP-4, P29C-ANCA antigen, PR-3CANCA, proteinase 3, serine proteinase, neutrophil, Wegener autoantigen, Wegener granulomatosis autoantigen | |
| Mouse | |
| 28 kDa | |
| 0.025 mL | |
| Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers | |
| 5657 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction