missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteinase 3/Myeloblastin/PRTN3 Antibody (3B4), Novus Biologicals™
Mouse Monoclonal Antibody
€ 205.00
Specifications
| Antigen | Proteinase 3/Myeloblastin/PRTN3 |
|---|---|
| Clone | 3B4 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, ELISA 1:10000 |
| Applications | Western Blot |
Description
Proteinase 3/Myeloblastin/PRTN3 Monoclonal specifically detects Proteinase 3/Myeloblastin/PRTN3 in Human samples. It is validated for Western Blot, ELISA.Specifications
| Proteinase 3/Myeloblastin/PRTN3 | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers | |
| ACPA, AGP7PR3, azurophil granule protein 7, C-ANCA, EC 3.4.21, EC 3.4.21.76, Leukocyte proteinase 3, MBN, MBT, myeloblastin, Neutrophil proteinase 4, NP4, NP-4, P29C-ANCA antigen, PR-3CANCA, proteinase 3, serine proteinase, neutrophil, Wegener autoantigen, Wegener granulomatosis autoantigen | |
| PRTN3 | |
| IgG2b | |
| Protein G purified | |
| 28 kDa |
| 3B4 | |
| Western Blot 1:500-1:2000, ELISA 1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 5657 | |
| Purified recombinant fragment of human Proteinase 3 expressed in E. Coli. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title