missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prothrombin Antibody (SR2163), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21771-100ul
This item is not returnable.
View return policy
Description
Prothrombin Monoclonal antibody specifically detects Prothrombin in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| Prothrombin | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| SR2163 | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1:50 | |
| Coagulation factor II, coagulation factor II (thrombin), EC 3.4.21, EC 3.4.21.5, prothrombin, prothrombin B-chain, PT, serine protease | |
| A synthesized peptide derived from human Prothrombin (Uniprot #: P00734) | |
| 100 μg | |
| Apoptosis, Breast Cancer, Cancer | |
| 2147 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction