missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prothrombin Antibody (SR2163), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 349.00 - € 472.00
Specifications
| Antigen | Prothrombin |
|---|---|
| Clone | SR2163 |
| Dilution | Western Blot 1:500-1:2000, Immunoprecipitation 1:50 |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18656124
|
Novus Biologicals
NBP3-21771-100ul |
100 μg |
€ 472.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18651284
|
Novus Biologicals
NBP3-21771-50ul |
50 μg |
€ 349.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Prothrombin Monoclonal antibody specifically detects Prothrombin in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
| Prothrombin | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Coagulation factor II, coagulation factor II (thrombin), EC 3.4.21, EC 3.4.21.5, prothrombin, prothrombin B-chain, PT, serine protease | |
| A synthesized peptide derived from human Prothrombin (Uniprot #: P00734) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR2163 | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Breast Cancer, Cancer | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 2147 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title