missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSP94/MSMB Antibody (SR1218), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21895-50ul
This item is not returnable.
View return policy
Description
PSP94/MSMB Monoclonal antibody specifically detects PSP94/MSMB in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation
Specifications
| PSP94/MSMB | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| SR1218 | |
| Western Blot 1:1000-1:5000, Immunohistochemistry 1:100-1:500, Immunoprecipitation 1:50 | |
| IGBF, MSMB, MSPB, PIP, PN44, PRPS, PSP57, PSP94 | |
| A synthesized peptide derived from human PSP94/MSMB (Uniprot #: P08118) | |
| 50 μg | |
| Cancer | |
| 4477 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction