missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSP94/MSMB Antibody (SR1218), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 338.00 - € 449.00
Specifications
| Antigen | PSP94/MSMB |
|---|---|
| Clone | SR1218 |
| Dilution | Western Blot 1:1000-1:5000, Immunohistochemistry 1:100-1:500, Immunoprecipitation 1:50 |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18629933
|
Novus Biologicals
NBP3-21895-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18613614
|
Novus Biologicals
NBP3-21895-50ul |
50 μg |
€ 338.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PSP94/MSMB Monoclonal antibody specifically detects PSP94/MSMB in Human samples. It is validated for Western Blot, Immunohistochemistry, ImmunoprecipitationSpecifications
| PSP94/MSMB | |
| Western Blot 1:1000-1:5000, Immunohistochemistry 1:100-1:500, Immunoprecipitation 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| IGBF, MSMB, MSPB, PIP, PN44, PRPS, PSP57, PSP94 | |
| A synthesized peptide derived from human PSP94/MSMB (Uniprot #: P08118) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1218 | |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Cancer | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 4477 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title