Learn More
Cowie Technology™ PTFE Joint Adapter
Shop All Fisherbrand Products
Click to view available options
Standard Taper Joint No.:
10/19
19/26
29/42
35/45
Cone Size:
14/23
24/29
29/32
45/50
Unit Size:
Each
Description
- Made from PTFE with a substantial shoulder for ease of handling
- Overall robust construction for repeated use
- Rigid sleeves and adapters are inert, usable to 280°C (approx.)
- Withstands high vacuum
- Transverse trapezoidal sealing rings on the contacting surfaces create high vacuum
Specifications
Specifications
| Cone Size | 45/50 |
| Product Type | PTFE Joint Adapters |
| Quantity | 1 Each |
| Standard Taper Joint No. | 29/42 |
Product Content Correction
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur