Learn More
Fisherbrand™ Right Angle Connectors
Product Code. 10212002 Shop All Fisherbrand Products
Change view
Click to view available options
Diameter (Metric) Inner:
10 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner unitSize
10212002 10 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10212002
Only null left
Add to basket
Add to basket
For use with rigid and semi-rigid tubing consisting of glass, FEP or PTFE plastic. Fisherbrand™ Right Angle Connectors feature all wetted parts manufactured from pure PTFE plastic.
  • Bayonet fitting
  • New, improved connector system
Order Info
dimProductType
Specifications
Certifications/Compliance Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
Autoclavable Autoclavable
Diameter (Metric) Inner 10 mm
Material PTFE
Overall Length (Metric) 34 mm
For Use With Flexible tubing in PTFE, Viton, silicone rubber or similar materials
Min. Operating Temperature (Metric) -200°C
Packaging Quantity 1 Each
Width (Metric) 33.5 mm
Height (Metric) 16 mm
Color White
Max. Operating Temperature (Metric) 280°C
Product Type PTFE Right Angle Connector