Learn More
JG Finneran Associates™ PTFE Seals
Description
- PTFE seals
- For use with dram sample vials
- Available in a variety of sizes
Specifications
Specifications
| Material (Liner) | PTFE |
| Color | White |
| Quantity | 1000 Pk. |
| Closure Size | 10 mm |