Learn More
Fisherbrand™ PTFE Test Tubes, Round Bottom with Lip
Product Code. 16275971 Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (Metric):
8 mL
13 mL
50 mL
85 mL
Diameter (Metric):
12 mm
16 mm
25 mm
40 mm
Length (Metric):
100 mm
145 mm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Diameter (Metric) Length (Metric) unitSize
16275971 85 mL 40 mm 100 mm Each
16245971 8 mL 12 mm 100 mm Each
16255971 13 mL 16 mm 100 mm Each
16265971 50 mL 25 mm 145 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16275971
Add to basket
Add to basket
Fisherbrand™ PTFE Test Tubes, Round Bottom with Lip have a smooth internal finish and are usable to 280°C.
  • Round Bottom with Lip
  • Total Chemical Resistance
  • High Purity
  • Temperature Range -200°C to +280°C
  • Non contaminating
  • Easy to clean
Specifications
Capacity (Metric) 85 mL
Material PTFE
Quantity 1
Product Line Fisherbrand
Length (Metric) 100 mm
Bottom Shape Round
Diameter (Metric) 40 mm
Product Type Round-bottom Test Tubes with Lip