Learn More
PTP gamma/PTPRG Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93896-0.02ml
This item is not returnable.
View return policy
Description
PTP gamma/PTPRG Polyclonal antibody specifically detects PTP gamma/PTPRG in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| PTP gamma/PTPRG | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| EC 3.1.3.48, H_RG317H01.1, HPTPG, protein tyrosine phosphatase, receptor type, G, protein tyrosine phosphatase, receptor type, gamma polypeptide, Protein-tyrosine phosphatase gamma, PTPGprotein tyrosine phosphatase gamma, receptor type protein tyrosine phosphatase gamma, receptor tyrosine phosphatase gamma, receptor-type protein phosphatase gamma, receptor-type tyrosine-protein phosphatase gamma, RPTPG, R-PTP-GAMMA | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 250-450 of human PTPRG (NP_002832.3). YRYTGSLTTPPCSEIVEWIVFRRPVPISYHQLEAFYSIFTTEQQDHVKSVEYLRNNFRPQQRLHDRVVSKSAVRDSWNHDMTDFLENPLGTEASKVCSSPPIHMKVQPLNQTALQVSWSQPETIYHPPIMNYMISYSWTKNEDEKEKTFTKDSDKDLKATISHVSPDSLYLFRVQAVCRNDMRSDFSQTMLFQANTTRIFQ | |
| 0.02 mL | |
| Signal Transduction | |
| 5793 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |