missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTP gamma/PTPRG Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 190.00 - € 470.00
Specifications
| Antigen | PTP gamma/PTPRG |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18637250
|
Novus Biologicals
NBP2-93896-0.02ml |
0.02 mL |
€ 190.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18645381
|
Novus Biologicals
NBP2-93896-0.1ml |
0.1 mL |
€ 470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PTP gamma/PTPRG Polyclonal antibody specifically detects PTP gamma/PTPRG in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| PTP gamma/PTPRG | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 5793 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| EC 3.1.3.48, H_RG317H01.1, HPTPG, protein tyrosine phosphatase, receptor type, G, protein tyrosine phosphatase, receptor type, gamma polypeptide, Protein-tyrosine phosphatase gamma, PTPGprotein tyrosine phosphatase gamma, receptor type protein tyrosine phosphatase gamma, receptor tyrosine phosphatase gamma, receptor-type protein phosphatase gamma, receptor-type tyrosine-protein phosphatase gamma, RPTPG, R-PTP-GAMMA | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 250-450 of human PTPRG (NP_002832.3). YRYTGSLTTPPCSEIVEWIVFRRPVPISYHQLEAFYSIFTTEQQDHVKSVEYLRNNFRPQQRLHDRVVSKSAVRDSWNHDMTDFLENPLGTEASKVCSSPPIHMKVQPLNQTALQVSWSQPETIYHPPIMNYMISYSWTKNEDEKEKTFTKDSDKDLKATISHVSPDSLYLFRVQAVCRNDMRSDFSQTMLFQANTTRIFQ | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title