missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Puratrophin-1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 196.00 - € 468.00
Specifications
| Antigen | Puratrophin-1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
Puratrophin-1 Polyclonal antibody specifically detects Puratrophin-1 in Rat samples. It is validated for Western BlotSpecifications
| Puratrophin-1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Neurodegeneration, Neuroscience | |
| PBS (pH 7.3), 50% glycerol | |
| 25894 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Rat | |
| ARHGEF44, DKFZP434I216, PH domain-containing family G member 4, pleckstrin homology domain containing, family G (with RhoGef domain) member 4, Pleckstrin homology domain-containing family G member 4, PRTPHN1, puratrophin-1, Purkinje cell atrophy associated protein 1, Purkinje cell atrophy-associated protein 1, SCA4, spinocerebellar ataxia 4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 710-1010 of human PLEKHG4 (NP_001123199.1). GALPQASPTVPPPGSSDPRSLNRLQLVLAEMVATEREYVRALEYTMENYFPELDRPDVPQGLRGQRAHLFGNLEKLRDFHCHFFLRELEACTRHPPRVAYAFLRHRVQFGMYALYSKNKPRSDALMSSYGHTFFKDKQQALGDHLDLASYLLKPIQRMGKYALLLQELARACGGPTQELSALREAQSLVHFQLRHGNDLLAMDAIQGCDVNLKEQGQLVRQDEFVVRTGRHKSVRRIFLFEELLLFSKPRHGPTGVDTFAYKRSFKMADLGLTECCGNSNLRFEIWFRRRKARDTFVLQAS | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title