Learn More
Honeywell KCL™ Pure Fit 395 Gloves
Product Code. 10505495
Click to view available options
Size:
10
11
Unit Size:
10 pairs
Each
Product Code. 10505495
Brand: Honeywell KCL™ 395/10
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Honeywell KCL™ Pure Fit 395 Gloves feature a foamed nitrile palm and fingertip coating.
  • Lightweight
  • Black nylon liner
  • Gray foamed nitrile palm and fingertip coating
  • Low linting
  • Comfortable
  • High tensile strength
  • High abrasion resistance
  • Provide dexterity and tactile sensitivity for precision work
  • Excellent grip in oily applications
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ISO 9001 / 2000
Coated Area Palm and fingertip
Coating Material Nitrile Foam
Color Gray, Black
Cut Resistance Rating 1
Size 10
Lining Lightweight black nylon
Quantity 144/Pk
Tear Rating 3