Learn More
Pyrex™ Borosilicate Glass Non-vacuum Desiccator Cover
Made of Borosilicate glass
Brand: Pyrex 1590/04D
119.70 EUR valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Additional Details : Weight : 0.72000kg
Description
Made of Borosilicate glass
Spare cover for non-vacuum desiccatorSpecifications
Spare Cover | |
for drying under atmospheric pressure | |
Flange: 170 mm |
Borosilicate Glass | |
Non-vacuum desiccator |