Learn More
Pyrrole-2-thiocarboxamide, 97%, Thermo Scientific™
Product Code. 15464408
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15464408 1 g 1g
15474408 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15464408 Supplier Thermo Scientific Alfa Aesar Supplier No. H58042.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 37488-45-2
Molecular Formula C5H6N2S
Molecular Weight (g/mol) 126.177
MDL Number MFCD00123524
InChI Key KJEMJZMJDZLKRH-UHFFFAOYSA-N
Synonym pyrrole-2-thiocarboxamide, pyrrole-2-carbothioamide, 1h-pyrrole-2-carbothioicacid;amide
PubChem CID 13589313
IUPAC Name 1H-pyrrole-2-carbothioamide
SMILES C1=CNC(=C1)C(=S)N
Specifications
Quantity 1 g
Formula Weight 126.18
Percent Purity 97%
Chemical Name or Material Pyrrole-2-thiocarboxamide