missing translation for 'onlineSavingsMsg'
Learn More
Learn More
QRSL Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94708-0.02ml
This item is not returnable.
View return policy
Description
QRSL Polyclonal antibody specifically detects QRSL in Human, Mouse samples. It is validated for Western Blot
Specifications
| QRSL | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| DKFZp564C1278, EC 6.3.5, EC 6.3.5.-, FLJ10989, FLJ12189, FLJ13447, GatA, glutaminyl-tRNA synthase (glutamine-hydrolyzing)-like 1, Glutaminyl-tRNA synthase-like protein 1, glutamyl-tRNA(Gln) amidotransferase subunit A homolog | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 289-528 of human QRSL1 (NP_060762.3). ELSSEVQSLWSKAADLFESEGAKVIEVSLPHTSYSIVCYHVLCTSEVASNMARFDGLQYGHRCDIDVSTEAMYAATRREGFNDVVRGRILSGNFFLLKENYENYFVKAQKVRRLIANDFVNAFNSGVDVLLTPTTLSEAVPYLEFIKEDNRTRSAQDDIFTQAVNMAGLPAVSIPVALSNQGLPIGLQFIGRAFCDQQLLTVAKWFEKQVQFPVIQLQELMDDCSAVLENEKLASVSLKQ | |
| 0.02 mL | |
| Endocrinology | |
| 55278 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction