missing translation for 'onlineSavingsMsg'
Learn More
Learn More
QRSL Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 196.00 - € 468.00
Specifications
| Antigen | QRSL |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
QRSL Polyclonal antibody specifically detects QRSL in Human, Mouse samples. It is validated for Western BlotSpecifications
| QRSL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Endocrinology | |
| PBS (pH 7.3), 50% glycerol | |
| 55278 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| DKFZp564C1278, EC 6.3.5, EC 6.3.5.-, FLJ10989, FLJ12189, FLJ13447, GatA, glutaminyl-tRNA synthase (glutamine-hydrolyzing)-like 1, Glutaminyl-tRNA synthase-like protein 1, glutamyl-tRNA(Gln) amidotransferase subunit A homolog | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 289-528 of human QRSL1 (NP_060762.3). ELSSEVQSLWSKAADLFESEGAKVIEVSLPHTSYSIVCYHVLCTSEVASNMARFDGLQYGHRCDIDVSTEAMYAATRREGFNDVVRGRILSGNFFLLKENYENYFVKAQKVRRLIANDFVNAFNSGVDVLLTPTTLSEAVPYLEFIKEDNRTRSAQDDIFTQAVNMAGLPAVSIPVALSNQGLPIGLQFIGRAFCDQQLLTVAKWFEKQVQFPVIQLQELMDDCSAVLENEKLASVSLKQ | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title