Learn More
Honeywell™ QUAD (i)XTREM S3 Shoes
Honeywell™ QUAD (i)XTREM S3 Shoes are constructed of water-resistant full grain leather.
Brand: Honeywell™ 6246212-40/7
Additional Details : Weight : 2.02000kg
Description
- Water resistant grain full leather
- Comfort foam for improved foot stability
- Gusset to avoid water and dust penetration
- Poromax 3D lining
- Toe cap metal free
- Metal free antiperforation insert
- Injected PU / Nitrile sole
- (i)NTELLIGENT wear control
- EN ISO 20345:2011 S3 HRO HI CI SRC
Specifications
EN ISO 20345:2011 S3 HRO HI CI SRC | |
Leather | |
Polyurethane and nitrile |
Brown, Black | |
40 | |
Metal-free |