Learn More
Socorex™ Qualitix™ Micropipette Tips, 1000μL, natural
Superior, autoclavable micropipette tips cover a range of sizes and fit a broad array of micropipette brands. Socorex™ Qualitix™ Micropipette Tips, 1000μL also come in a variety of shapes to facilitate fluid transfer.
Brand:  Socorex™ 3070.1000RS
Additional Details : Weight : 0.41000kg
 View more versions of this product
Product Code. 15740659
  • € 86.40 / Pack of 576
Estimated Shipment: 18-07-2024
to see stock.
Add to basket
Description
Description
  • Cover a range of sizes.
  • Autoclavable.
  • Optimal tightness fitting on Socorex™ pipettes and broad compatibility with other brands.
  • Various tip versions available.
Specifications
Specifications
1000 μL
Non-filtered
Natural
Fine Point
6/PK, 8PK/CS
Universal Fit Micro
1000μL
Sterile
Autoclavable
96/Rack
Single-use
Qualitix