Learn More
Socorex™ Qualitix™ Micropipette Tips, 200μL
Superior, autoclavable micropipette tips cover a range of sizes and fit a broad array of micropipette brands. Socorex™ Qualitix™ Micropipette Tips, 200μL also come in a variety of shapes to facilitate fluid transfer.
Brand:  Socorex™ 3070.0200R
Additional Details : Weight : 0.78000kg
 View more versions of this product
Product Code. 15740649
  • € 69.80 / Pack of 960
Estimated Shipment: 03-06-2024
to see stock.
Add to basket
Description
Description
  • Cover a range of sizes.
  • Autoclavable.
  • Optimal tightness fitting on Socorex™ pipettes and broad compatibility with other brands.
  • Various tip versions available.
Specifications
Specifications
200 μL
Non-filtered
Natural
Beveled
10/PK, 10PK/CS
Universal Fit Micro
200μL
Non-sterile
Autoclavable
96/Rack
Single-use
Qualitix