Learn More
(R)-(-)-1,3-Butanediol, 98%
Product Code. 11446167
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11446167 1 g 1g
11456167 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11446167 Supplier Thermo Scientific Alfa Aesar Supplier No. B25124.03
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
CAS: 6290-03-5 | C8H15NO4 | 189.21 g/mol
(R)-(-)-1,3-Butanediol is used as a solvent for food flavoring agents and is a co-monomer used in certain polyurethane and polyester resins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(R)-(-)-1,3-Butanediol is used as a solvent for food flavoring agents and is a co-monomer used in certain polyurethane and polyester resins.
Solubility
Fully miscible in water.
Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, water/ moisture.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6290-03-5
MDL Number MFCD00064277
Specifications
Density 1.005
Boiling Point 203°C to 204°C
Flash Point 121°C (249°F)
Odor Weak
Molecular Formula C8H15NO4
Refractive Index 1.44
Quantity 1 g
Beilstein 1718944
Sensitivity Hygroscopic
Synonym diethyl iminodiacetate, diethyl 2,2'-azanediyldiacetate, diethyliminodiacetate, iminodiacetic acid diethyl ester, glycine, n-2-ethoxy-2-oxoethyl-, ethyl ester, diethyl 2,2'-iminodiacetate, n,n-bis ethoxycarbonylmethyl amine, acetic acid, iminodi-, diethyl ester, ethyl 2-2-ethoxy-2-oxoethyl amino acetate, zlchem 799
Solubility Information Fully miscible in water.
InChI Key LJDNMOCAQVXVKY-UHFFFAOYSA-N
Optical Rotation -30° (Neat)
SMILES CCOC(=O)CNCC(=O)OCC
IUPAC Name ethyl 2-[(2-ethoxy-2-oxoethyl)amino]acetate
Molecular Weight (g/mol) 189.21
PubChem CID 80502
Formula Weight 90.12
Percent Purity 98%
Chemical Name or Material (R)-(-)-1,3-Butanediol