missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB5B Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27403-1-AP-20UL
This item is not returnable.
View return policy
Description
RAB5B play a role in protein transport. RAB5B is probably involved in vesicular traffic.Specifications
| RAB5B | |
| Polyclonal | |
| Unconjugated | |
| RAB5B | |
| RAB5B, Ras related protein Rab 5B | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 19344, 5869 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunocytochemistry, Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P61020, P61021 | |
| RAB5B | |
| RAB5B Fusion Protein Ag26584 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction