missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB5B Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | RAB5B |
|---|---|
| Concentration | 0.3 mg/mL |
| Applications | Immunofluorescence, Immunocytochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16865605
|
Proteintech
27403-1-AP-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16855605
|
Proteintech
27403-1-AP-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RAB5B play a role in protein transport. RAB5B is probably involved in vesicular traffic.Specifications
| RAB5B | |
| Immunofluorescence, Immunocytochemistry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse, Human | |
| P61020, P61021 | |
| 19344, 5869 | |
| RAB5B Fusion Protein Ag26584 | |
| Primary | |
| -20°C | |
| RAB5B |
| 0.3 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| RAB5B, Ras related protein Rab 5B | |
| RAB5B | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title