missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RABGAP1L Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94545-0.02ml
This item is not returnable.
View return policy
Description
RABGAP1L Polyclonal antibody specifically detects RABGAP1L in Mouse samples. It is validated for Western Blot
Specifications
| RABGAP1L | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| DKFZp686E1450, FLJ58037, FLJ79118, heart, liver (HLL), HHLFLJ79254, RAB GTPase activating protein 1-like, rab GTPase-activating protein 1-like, TBC1 domain family, member 18, TBC1D18 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 50-210 of human RABGAP1L (NP_055672.3). NGDEQLEKAMEEILRDSEKRPSSLLVDCQSSSEISDHSFGDIPASQTNKPSLQLILDPSNTEISTPRPSSPGGLPEEDSVLFNKLTYLGCMKVSSPRNEVEALRAMATMKSSSQYPFPVTLYVPNVPEGSVRIIDQSSNVEIASFPIYKVLFCARGHDGTT | |
| 0.02 mL | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 9910 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction