Learn More
RAP1A Antibody (RAP1 4C8/1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-50269
 View more versions of this product
Product Code. 18677508
  • € 351.00 / 0.10mL
Description
RAP1A Monoclonal antibody specifically detects RAP1A in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RAP1A
Monoclonal
0.43 mg/mL
Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, CyTOF-ready
A2ALS5
Mouse
Protein A purified
RUO
Primary
Human, Mouse
Purified
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence
RAP1 4C8/1
Unconjugated
PBS
C21KG, G-22K, GTP-binding protein smg p21A, KREV-1, KREV1Ras-related protein Krev-1, RAP1, RAP1A, member of RAS oncogene family, ras-related protein Rap-1A, RAS-related protein RAP1A, SMGP21
Human RAP1A
0.1 mL
Cell Cycle and Replication, Endosome Markers
5906
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG2b