Learn More
RASSF1 Monoclonal Antibody (OTI2A9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA502481
Encompass_Preferred
 View more versions of this product
Product Code. 17577001
  • € 540.87 / 100µL
Description
This gene encodes a protein similar to the RAS effector proteins. Loss or altered expression of this gene has been associated with the pathogenesis of a variety of cancers, which suggests the tumor suppressor function of this gene. The inactivation of this gene was found to be correlated with the hypermethylation of its CpG-island promoter region. The encoded protein was found to interact with DNA repair protein XPA. The protein was also shown to inhibit the accumulation of cyclin D1, and thus induce cell cycle arrest. Seven alternatively spliced transcript variants of this gene encoding distinct isoforms have been reported.
Specifications
Specifications
RASSF1
Monoclonal
0.52 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9NS23
RASSF1
Full length human recombit protein of human RASSF1 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
OTI2A9
Unconjugated
RASSF1
IGKV1/OR-2, IGKV1OR2-PENDING
Mouse
Affinity Chromatography
RUO
11186
-20° C, Avoid Freeze/Thaw Cycles