missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RASSF1 Monoclonal Antibody (OTI2A9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
POA - € 242.00
Specifications
| Antigen | RASSF1 |
|---|---|
| Clone | OTI2A9 |
| Concentration | 0.52 mg/mL |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17597001
|
Origene Technologies
TA502481S |
30 μL |
€ 242.00
30µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17577001
|
Origene Technologies
TA502481 |
100 μL |
POA
100µL |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Beskrivelse
RASSF1 Monoclonal antibody specifically detects RASSF1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotTekniske data
| RASSF1 | |
| 0.52 mg/mL | |
| Unconjugated | |
| Mouse | |
| Human | |
| 11186 | |
| Primary | |
| RASSF1 |
| OTI2A9 | |
| Monoclonal | |
| Liquid | |
| RUO | |
| Q9NS23 | |
| IgG1 | |
| -20° C, Avoid Freeze/Thaw Cycles |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Fisher Scientific Encompass-programmet tilbyder varer, som ikke er en del af vores distributionsportefølje. Disse produkter har typisk ikke billeder eller detaljerede beskrivelser. Vi er dog forpligtet til at forbedre din shoppingoplevelse. Brug formularen nedenfor til at give feedback relateret til indholdet på dette produkt.
Produkttitel