missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBM4 Mouse anti-Human, Clone: 3H3F12, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60292-1-IG-150UL
This item is not returnable.
View return policy
Description
May play a role in alternative splice site selection during pre-mRNA processing.Specifications
| RBM4 | |
| Monoclonal | |
| 1.45 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9BWF3 | |
| RBM4 | |
| RBM4 Fusion Protein Ag2190 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 3H3F12 | |
| Unconjugated | |
| RBM4 | |
| hLark, LARK, Lark homolog, RBM4, RBM4A, RNA binding motif protein 4, RNA binding motif protein 4a, RNA binding protein 4, ZCCHC21, ZCRB3A | |
| Mouse | |
| Protein A | |
| RUO | |
| 5936 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction