missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBM4 Mouse anti-Human, Clone: 3H3F12, Proteintech
Mouse Monoclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | RBM4 |
|---|---|
| Clone | 3H3F12 |
| Concentration | 1.45 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16867365
|
Proteintech
60292-1-IG-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16857365
|
Proteintech
60292-1-IG-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
May play a role in alternative splice site selection during pre-mRNA processing.Specifications
| RBM4 | |
| 1.45 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| hLark, LARK, Lark homolog, RBM4, RBM4A, RNA binding motif protein 4, RNA binding motif protein 4a, RNA binding protein 4, ZCCHC21, ZCRB3A | |
| RBM4 | |
| IgG2a | |
| Protein A | |
| Antibody |
| 3H3F12 | |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9BWF3 | |
| 5936 | |
| RBM4 Fusion Protein Ag2190 | |
| Primary | |
| -20°C | |
| RBM4 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title