missing translation for 'onlineSavingsMsg'
Learn More

RBM4 Mouse anti-Human, Clone: 3H3F12, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 60292-1-IG-20UL

 View more versions of this product

Product Code. 16867365

  • € 145.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

May play a role in alternative splice site selection during pre-mRNA processing.
Specifications

Specifications

RBM4
Monoclonal
1.45 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9BWF3
RBM4
RBM4 Fusion Protein Ag2190
20 μL
Primary
Human
Antibody
IgG2a
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunofluorescence
3H3F12
Unconjugated
RBM4
hLark, LARK, Lark homolog, RBM4, RBM4A, RNA binding motif protein 4, RNA binding motif protein 4a, RNA binding protein 4, ZCCHC21, ZCRB3A
Mouse
Protein A
RUO
5936
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.