missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bovine Pregnancy-associated glycoprotein 1

A DNA sequence encoding the Bos taurus (Bovine) Pregnancy-associated glycoprotein 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7639-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16039960

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_776836.1
Pregnancy-associated glycoprotein 1
Research Use Only
PAG1
Recombinant Protein
E. coli
RGSNLTTHPL RNIKDLVYMG NITIGTPPQE FQVVFDTASS DLWVPSDFCT SPACSTHVRF RHLQSSTFRL TNKTFRITYG SGRMKGVVVH DTVRIGNLVS TDQPFGLSIE EYGFEGRIYD GVLGLNYPNI SFSGAIPIFD KLKNQRAISE PVFAFYLSKD EREGSVVMFG GVDHRYYEGE LNWVPLIQAG DWSVHMDRIS IERKIIACSD GCKALVDTGT SDIVGPRRLV NNIHRLIGAI PRGSEHYVPC SEVNTLPSIV FTINGINYPV PGRAYILKDD RGRCYTTFQE NRVSSSTETW YLGDVFLRLY FSVFDRGNDR IGLARAV
Greater than 88.1% as determined by SDS-PAGE.
281964
50 μg
Not determined.
Full Length of Mature Protein
Bovine
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers