missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine SNRPD Protein
A cDNA sequence encoding the SNRPD was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13546-1mg
Additional Details : Weight : 0.01000kg
Specifications
SNRPD Protein | |
Research Use Only | |
Recombinant Protein | |
Bovine | |
SNRPD is supplied in 20mM HEPES buffer pH 7.6, 250mM NaCl and 20% glycerol. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Bovine | |
Untagged | |
Greater than 60% as determined by SDS-PAGE. |