missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Dog Arginine esterase Protein
A DNA sequence encoding the Canis lupus familiaris (Dog) (Canis familiaris) Arginine esterase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7377-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001003284.1 | |
Arginine esterase Protein | |
Research Use Only | |
KLK2 | |
Recombinant Protein | |
E. coli | |
IIGGRECLKN SQPWQVAVYH NGEFACGGVL VNPEWVLTAA HCANSNCEVW LGRHNLSESE DEGQLVQVRK SFIHPLYKTK VPRAVIRPGE DRSHDLMLLH LEEPAKITKA VRVMDLPKKE PPLGSTCYVS GWGSTDPETI FHPGSLQCVD LKLLSNNQCA KVYTQKVTKF MLCAGVLEGK KDTCKGDSGG PLICDGELVG ITSWGATPCG KPQMPSLYTR VMPHLMWIKD TMKANT | |
Greater than 90% as determined by SDS-PAGE. |
403967 | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Dog | |
His-SUMO | |
Tris-based buffer, 50% glycerol |