missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Dog IL4 / Interleukin-4 Protein

A DNA sequence encoding the Canis lupus familiaris (Dog) (Canis familiaris) IL4 / Interleukin-4, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6222-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16021700

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001003159.1
IL4 / Interleukin-4 Protein
Research Use Only
IL4
Recombinant Protein
E. coli
HNFNITIKEI IKMLNILTAR NDSCMELTVK DVFTAPKNTS DKEIFCRAAT VLRQIYTHNC SNRYLRGLYR NLSSMANKTC SMNEIKKSTL KDFLERLKVI MQKKYYRH
Greater than 90% as determined by SDS-PAGE.
403785
10 μg
Not determined.
Full Length
Dog
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers