missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Dog IL4 / Interleukin-4 Protein
A DNA sequence encoding the Canis lupus familiaris (Dog) (Canis familiaris) IL4 / Interleukin-4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6222-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001003159.1 | |
IL4 / Interleukin-4 Protein | |
Research Use Only | |
IL4 | |
Recombinant Protein | |
E. coli | |
HNFNITIKEI IKMLNILTAR NDSCMELTVK DVFTAPKNTS DKEIFCRAAT VLRQIYTHNC SNRYLRGLYR NLSSMANKTC SMNEIKKSTL KDFLERLKVI MQKKYYRH | |
Greater than 90% as determined by SDS-PAGE. |
403785 | |
200 μg | |
Not determined. | |
Full Length | |
Dog | |
His | |
Tris-based buffer, 50% glycerol |