missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Horse Major allergen Equ c 1 Protein
A DNA sequence encoding the Equus caballus (Horse) Major allergen Equ c 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7892-ye-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001075966.1 | |
Major allergen Equ c 1 Protein | |
Research Use Only | |
LOC100034197 | |
Recombinant Protein | |
Yeast | |
QQEENSDVAI RNFDISKISG EWYSIFLASD VKEKIEENGS MRVFVDVIRA LDNSSLYAEY QTKVNGECTE FPMVFDKTEE DGVYSLNYDG YNVFRISEFE NDEHIILYLV NFDKDRPFQL FEFYAREPDV SPEIKEEFVK IVQKRGIVKE NIIDLTKIDR CFQLRGNGVA QA | |
Greater than 90% as determined by SDS-PAGE. |
100034197 | |
50 μg | |
Not determined. | |
Full Length | |
Horse | |
His | |
Tris-based buffer, 50% glycerol |