missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ABHD14B Protein
A DNA sequence encoding the Homo sapiens (Human) ABHD14B, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7914-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001139786.1 | |
ABHD14B Protein | |
Research Use Only | |
ABHD14B | |
Recombinant Protein | |
E. coli | |
AASVEQREGT IQVQGQALFF REALPGSGQA RFSVLLLHGI RFSSETWQNL GTLHRLAQAG YRAVAIDLPG LGHSKEAAAP APIGELAPGS FLAAVVDALE LGPPVVISPS LSGMYSLPFL TAPGSQLPGF VPVAPICTDK INAANYASVK TPALIVYGDQ DPMGQTSFEH LKQLPNHRVL IMKGAGHPCY LDKPEEWHTG LLDFLQGLQ | |
Greater than 90% as determined by SDS-PAGE. |
84836 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |