missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Beta-defensin 124 Protein
A DNA sequence encoding the Homo sapiens (Human) Beta-defensin 124, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7883-ye-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001032589.1 | |
Beta-defensin 124 Protein | |
Research Use Only | |
DEFB124 | |
Recombinant Protein | |
Yeast | |
EFKRCWKGQG ACQTYCTRQE TYMHLCPDAS LCCLSYALKP PPVPKHEYE | |
Greater than 90% as determined by SDS-PAGE. |
245937 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |