missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human BPHL Protein

A DNA sequence encoding the Homo sapiens (Human) BPHL, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7794-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16010841

  • € 2385.00 / 1mg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001289706.1
BPHL Protein
Research Use Only
BPHL
Recombinant Protein
E. coli
MPRNLLYSLL SSHLSPHFST SVTSAKVAVN GVQLHYQQTG EGDHAVLLLP GMLGSGETDF GPQLKNLNKK LFTVVAWDPR GYGHSRPPDR DFPADFFERD AKDAVDLMKA LKFKKVSLLG WSDGGITALI AAAKYPSYIH KMVIWGANAY VTDEDSMIYE GIRDVSKWSE RTRKPLEALY GYDYFARTCE KWVDGIRQFK HLPDGNICRH LLPRVQCPAL IVHGEKDPLV PRFHADFIHK HVKGSRLHLM PEGKHNLHLR FADEFNKLAE DFLQ
Greater than 90% as determined by SDS-PAGE.
670
1 mg
Not determined.
Full Length of Isoform 2
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers